Sulfuric acid 14% (v/v) in aqueous solution

Supplier: Ricca Chemical

R8160000-500A R8160000-1A
RC816016EA 56.88 CAD
RC816016 RC8160-32
Sulfuric acid 14% (v/v) in aqueous solution
Sulfuric acid
Formula: H₂SO₄
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 1830
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 14.00 ± 0.03 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR