5-Sulfosalicylic acid 5% (w/v) in aqueous solution
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
2-Hydroxy-5-sulphobenzoic acid, 2-Hydroxy-5-sulfobenzoic acid
R8120000-500C
R8120000-1C
RC812016EA
102.7
CAD
RC812016
RC812032
5-Sulfosalicylic acid 5% (w/v) in aqueous solution
5-Sulfosalicylic acid solutions
Formula:
HO₃SC₆H₃-2-(OH)CO₂H·2H₂O Boiling Pt: ~ 100 °C Melting Pt: ~ 0 °C Density: 1.0300 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00007508 CAS Number: 5965-83-3 |
Specification Test Results
Appearance | Clear, slight tannish gold to slight pink, odorless |
Assay at 20 °C (traceable to NIST SRM 84) | 5.00 ± 0.02 % (w/v) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...