Sulfanilamide reagent 1% (w/v) in hydrochloric acid 10%

Supplier: Ricca Chemical
Danger

Synonyms: 4-Aminobenzenesulfonamide, 4-Aminobenzenesulphonamide

R8103000-500A R8103000-1A
RC810316EA 143.78 CAD
RC810316 RC8103-32
Sulfanilamide reagent 1% (w/v) in hydrochloric acid 10%
Sulphanilamide
Formula: H₂NC₆H₄SO₂NH₂
MW: 172.21 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0200 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00007939
CAS Number: 63-74-1
UN: 1789
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR