Crystal violet in acetic acid (Spinal diluting fluid)

Supplier: Ricca Chemical
Danger

R7996000-500A R7996000-120A
RC799616EA 126.4 CAD
RC799616 RC79964
Crystal violet in acetic acid (Spinal diluting fluid)
Crystal Violet

Crystal Violet-Acetic Acid Solution for Cerebrospinal Fluid Cell Count

Formula: C₂₅H₃₀ClN₃
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0100 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011750
CAS Number: 548-62-9
UN: 2790
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, purple, Acetic odor


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR