Sodium thiosulfate 0.0375 N

Supplier: Ricca Chemical
Warning

R7925000-4A R7925000-1A
RC79251EA 232.26 CAD
RC79251 RC792532
Sodium thiosulfate 0.0375 N
Sodium thiosulphate
Formula: Na₂S₂O₃
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0050 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7

Order Now

Specification Test Results

Appearance Clear, colorless, slight organic odor
Assay at 20 °C (traceable to NIST SRM 136) 0.03750 ± 0.00001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR