Sodium thiosulfate 0.0250 N (N/40)

Supplier: Ricca Chemical
Warning

Synonyms: di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite

R7900000-4A R7900000-1A
RC79001EA 229.1 CAD
RC79001 RC790032
Sodium thiosulfate 0.0250 N (N/40)
Sodium thiosulphate
Formula: Na₂S₂O₃
MW: 158.11 g/mol
Boiling Pt: 100
Melting Pt: 48
Density: 1.0040 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
Merck Index: 12,08844

Order Now

Specification Test Results

Appearance Clear, colorless, slight organic odor
Assay at 20 °C (traceable to NIST SRM 136) 0.02500 ± 0.00001 N at 20.0 °C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR