Disodium oxalate 0.100 M (0.200 N)

Supplier: Ricca Chemical

R7500000-500A R7500000-120A
RC750016EA 94.8 CAD
RC750016 RC75004
Disodium oxalate 0.100 M (0.200 N)
di-Sodium oxalate
Formula: Na₂C₂O₄
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0010 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00012465
CAS Number: 62-76-0

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 40) 0.1000 ± 0.0001 M at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR