Sodium nitroprusside 0.5 g/L in aqueous solution

Supplier: Ricca Chemical

R7498000-500C R7498000-1C
RC7498-16EA 97.96 CAD
RC7498-16 RC7498-32
Sodium nitroprusside 0.5 g/L in aqueous solution
Sodium nitroprusside
Formula: C₅FeN₆Na₂O
MDL Number: MFCD00003506
CAS Number: 14402-89-2
UN: 1588
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear, tan color, odorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR