Sodium nitrite 5% (w/w) in aqueous solution

Supplier: Ricca Chemical

Synonyms: Diazotizing salts

R7497000-500C R7497000-1C
RC7497-32EA 155.11 CAD
RC7497-32 RC7497-16
Sodium nitrite 5% (w/w) in aqueous solution
Sodium nitrite
Formula: NaNO₂
MW: 69 g/mol
MDL Number: MFCD00011118
CAS Number: 7632-00-0
UN: 1500
ADR: 5.1,III

Order Now

Specification Test Results

Appearance Clear, colorless to slight yellow, odorless
Refractive Index Confirmed Concentration

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR