Sodium hydroxide : sodium thiosulfate (50:2.5) % (w/v) in aqueous solution for Kjeldahl nitrogen analysis using copper catalyst
Supplier: Ricca Chemical
|
Danger
|
R7495000-4A
R7495000-10F
R7495000-500A
RC74951EA
206.98
CAD
RC74951
RC749525
RCR7495000500A
Sodium hydroxide : sodium thiosulfate (50:2.5) % (w/v) in aqueous solution for Kjeldahl nitrogen analysis using copper catalyst
Sodium hydroxide : sodium thiosulfate
Boiling Pt:
~ 100 °C Density: 1.4000 g/cm³ Storage Temperature: Ambient |
UN:
1824 ADR: 8,II |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 136) | 2.5 ± 0.1 % (w/v) |
Assay at 20 °C (traceable to NIST SRM 723) | 50.0 ± 0.4 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...