Sodium hydroxide 0.100 N (N/10) in methanol

Supplier: Ricca Chemical
Danger

Synonyms: Caustic soda

R7472000-4A R7472000-1A R7472000-20F
RC7472-1CS 1817 CAD
RC7472-1 RCR7472000-1A RCR7472000-20F
Sodium hydroxide 0.100 N (N/10) in methanol
Sodium hydroxide
Formula: NaOH
Melting Pt: –98°C
Density: 0.8
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 0.0995-0.1005 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR