Sodium hydroxide 0.1562 N
Supplier: Ricca Chemical
R7357000-4A
R7357000-1A
R7357000-20F
RC7357-1EA
164.32
CAD
RC7357-1
RC7357-32
RC7357-5
Sodium hydroxide 0.1562 N
Sodium hydroxide
Formula:
NaOH |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 UN: 1823 ADR: 8,II |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 84) | 0.1562 ± 0.0001 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...