Sodium hydroxide 0.0227 N
Supplier: Ricca Chemical
Synonyms:
Caustic soda
R7310000-4A
R7310000-1A
RC73101EA
192.76
CAD
RC73101
RC731032
Sodium hydroxide 0.0227 N
Sodium hydroxide
1 mL = 1 mg CO2
Formula:
NaOH MW: 40 g/mol |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 UN: 1823 ADR: 8,II |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 84) | 0.02270 ± 0.00005 N at 20.0 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...