Sodium hydroxide 8% (w/v) in aqueous solution

Supplier: Ricca Chemical

R7256000-500A R7256000-1A
RC725616EA 97.96 CAD
RC725616 RC7256-32
Sodium hydroxide 8% (w/v) in aqueous solution
Sodium hydroxide
Formula: NaOH
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 8.00 ± 0.04 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR