tri-Sodium citrate 3.8% (w/v) in aqueous solution

Supplier: Ricca Chemical

R7228000-500A R7228000-120A
RC722816EA 112.18 CAD
RC722816 RC72284
tri-Sodium citrate 3.8% (w/v) in aqueous solution
tri-Sodium citrate solutions
Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0250 g/cm³
MDL Number: MFCD00150031
CAS Number: 6132-04-3

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay 3.8 ± 0.3 % w/v

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR