Sodium chloride 0.0250 N (N/40)
Supplier: Ricca Chemical
R7222000-1A
R7222000-500A
RC722232EA
121.66
CAD
RC722232
RC7222-16
Sodium chloride 0.0250 N (N/40)
Sodium chloride
Formula:
NaCl Boiling Pt: 100 °C Melting Pt: 0.0 °C Density: 1.0011 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 999) | 0.02500 ± 0.00001 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...