Sodium chloride 30% (w/v) in aqueous solution

Supplier: Ricca Chemical

R7220000-1A R7220000-500A
RC7220-32EA 251.22 CAD
RC7220-32 RC722016
Sodium chloride 30% (w/v) in aqueous solution
Sodium chloride
Formula: NaCl
Boiling Pt: ~ 100 °C
Melting Pt: ~ –10
Density: 1.1925 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay 30.0 ± 0.3 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR