Sodium chloride 10% (w/v) in aqueous solution

Supplier: Ricca Chemical

R7217000-500A R7217000-1A
RC7217-16EA 412.38 CAD
RC7217-16 RC7217-32
Sodium chloride 10% (w/v) in aqueous solution
Sodium chloride
Formula: NaCl
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0680 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Refractive Index 10.0 ± 0.2 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR