Sodium carbonate 0.0500 N (N/20)
Supplier: Ricca Chemical
Synonyms:
Calcined soda, Carbonic acid disodium salt
R7185000-500A
R7185000-1A
RC718516EA
90.06
CAD
RC718516
RC718532
Sodium carbonate 0.0500 N (N/20)
Sodium carbonate
1 ml = 2.5 mg CaCO₃
Formula:
Na₂CO₃ MW: 105.99 g/mol Boiling Pt: 1600 Melting Pt: 854 Density: 1.0028 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003494 CAS Number: 497-19-8 Merck Index: 14,08596 |
|
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 723) | 0.05000 ± 0.00001 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...