Sodium carbonate 0.1% (w/v) in aqueous solution

Supplier: Ricca Chemical

R7150000-4A R7150000-500A R7150000-1A
RC71501EA 252.8 CAD
RC71501 RC715016 RC715032
Sodium carbonate 0.1% (w/v) in aqueous solution
Sodium carbonate
Formula: Na₂CO₃
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0010 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 0.1000 ± 0.0003 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR