Simulated gastric fluid (without pepsin) (0,2% NaCl) (w/v) in hydrochloric acid 0.7%
Supplier: Ricca Chemical
|
Danger
|
R7108000-10F
R7108000-1A
R7108000-20F
RC7108-2.5EA
984.34
CAD
RC7108-2.5
RC7108-32
RC7108-5
Simulated gastric fluid (without pepsin) (0,2% NaCl) (w/v) in hydrochloric acid 0.7%
Simulated gastric fluid (without pepsin)
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 999) | 0.1188 ± 0.0050 N at 20.0 °C |
pH at 25 °C (traceable to NIST SRM 185 & 186) | 1.2 ± 0.2 pH at 25.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...