Ammonium thiocyanate 30% w/v aqueous solution

Supplier: Ricca Chemical
Danger

Synonyms: Thiocyanic acid ammonium salt, Ammonium rhodanide, Ammonium sulphocyanate, Ammonium sulfocyanate

R0698000-4A R0698000-1A
RC698-1EA 609.88 CAD
RC698-1 RC698-32
Ammonium thiocyanate 30% w/v aqueous solution
Ammonium thiocyanate
Formula: NH₄SCN
MW: 76.12 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: 150
Density: 1.1000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011428
CAS Number: 1762-95-4
Merck Index: 13,00564

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Suitability for Use Red color with Iron (Fe)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR