Silver nitrate 0.0171 N
Supplier: Ricca Chemical
R6900000-4B
R6900000-1B
R6900000-20F
RC69001EA
338.12
CAD
RC69001
RC690032
RC69005
Silver nitrate 0.0171 N
Silver nitrate
1 mL = 1 mg NaCl
Formula:
AgNO₃ |
MDL Number:
MFCD00003414 CAS Number: 7761-88-8 UN: 1493 ADR: 5.1,II |
|
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 999) | 0.01710 ± 0.00001 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...