Ammonium heptamolybdate 5% (w/v) in aqueous solution USP Reference Solution (RS)

Supplier: Ricca Chemical
Warning

R0675000-500A R0675000-120A
RC675-16EA 276.5 CAD
RC675-16 RC675-4
Ammonium heptamolybdate 5% (w/v) in aqueous solution USP Reference Solution (RS)
Ammonium heptamolybdate solutions
Formula: (NH₄)₆Mo₇O₂₄·4H₂O
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2

Order Now

Specification Test Results

Appearance Clear, colorless, odorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR