Ammonium heptamolybdate 10% (w/v) in aqueous solution, pH adjusted

Supplier: Ricca Chemical
Danger

R0670000-500A R0670000-1A
RC67016EA 233.84 CAD
RC67016 RC670-32
Ammonium heptamolybdate 10% (w/v) in aqueous solution, pH adjusted
Ammonium heptamolybdate solutions
Formula: (NH₄)₆Mo₇O₂₄·4H₂O
Boiling Pt: ~ 100 °C
Density: 1.1000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2

Order Now

Specification Test Results

Appearance Clear, colorless liquid
pH at 25 °C (traceable to NIST SRM 186 & 191) 7.5 ± 0.5 pH at 25.0 C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR