Safranine O alcoholic solution counterstain for gram staining

Supplier: Ricca Chemical
Danger

Synonyms: Gossypimine, Safranin O, Safranin Y, Safranin A, 3,7-Diamino-2,8-dimethyl-5-phenylphenazinium chloride, 3,7-Diamino-2,8-dimethyl-5-phenylphenazin-5-ium chloride, Safranin T, CI 50240, Basic Red 2, Safranin, Cotton Red

R6680000-500A R6680000-1A
RC668016EA 121.66 CAD
RC668016 RC668032
Safranine O alcoholic solution counterstain for gram staining
Safranine O
Formula: C₂₀H₁₉ClN₄
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 0.9000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011759
CAS Number: 477-73-6

Order Now

Specification Test Results

Absorbance (traceable to NIST SRM 930) Conforms to control
Appearance Clear, dark red, slight alcohol odor

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR