Safranine O 1% (w/v) in aqueous solution counterstain for gram staining

Supplier: Ricca Chemical

R6660000-500A R6660000-120A
RC666016EA 124.82 CAD
RC666016 RC66604
Safranine O 1% (w/v) in aqueous solution counterstain for gram staining
Safranine O
Formula: C₂₀H₁₉ClN₄
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0100 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011759
CAS Number: 477-73-6

Order Now

Specification Test Results

Absorbance (traceable to NIST SRM 930) Conforms to control
Appearance Dark red, odorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR