Robert's reagent for the qualitative detection of albumin in urine
Supplier: Ricca Chemical
|
Danger
|
R6630000-500C
R6630000-1C
RC663016EA
134.3
CAD
RC663016
RC663032
Robert's reagent for the qualitative detection of albumin in urine
Robert's reagent
Boiling Pt:
~ 102 °C Density: 1.2800 g/cm³ Storage Temperature: Ambient |
UN:
2031 ADR: 8,II |
Specification Test Results
Appearance | Clear, colorless, odorless |
Refractive Index | Confirms correct formulation |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...