Potassium thiocyanate 4% (w/v) in aqueous solution
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Potassium rhodanide
R6550000-500A
R6550000-1A
RC6550-16EA
102.7
CAD
RC6550-16
RC6550-32
Potassium thiocyanate 4% (w/v) in aqueous solution
Potassium thiocyanate
Formula:
KSCN MW: 97.18 g/mol Boiling Pt: 500 Melting Pt: 175 Density: 1.0195 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011413 CAS Number: 333-20-0 Merck Index: 12,07866 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 999) | 4.00 ± 0.05 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...