Potassium pyrogallate for Orsat gas analysis
Supplier: Ricca Chemical
|
Danger
|
R6460000-500B
R6460000-1B
RC646016EA
320.74
CAD
RC646016
RC646032
Potassium pyrogallate for Orsat gas analysis
Potassium pyrogallate
Formula:
C₆H₃K₃O₃ Boiling Pt: ~ 102 °C Density: 1.3741 g/cm³ Storage Temperature: Ambient |
UN:
1814 ADR: 8,II |
Specification Test Results
Appearance | Dark brown, odorless |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...