Ammonia solution 50% (v/v)

Supplier: Ricca Chemical
Danger

R0640000-500A R0640000-1A R0640000-16E R0640000-20F R0640000-200E
RC640-16EA 132.72 CAD
RC640-16 RC640-32 RCR0640000-16E RCR0640000-20F RCR0640000200E
Ammonia solution 50% (v/v)
Ammonia solution
Formula: NH₃
Boiling Pt: ~ 36 °C
Melting Pt: ~ 0 °C
Density: 0.9500 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Methyl Red) 48.5 - 51.5 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR