Ammonia solution 20%
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
R0635000-10F
R0635000-20F
R0635000-4A
RCR0635000-10FEA
511.92
CAD
RCR0635000-10F
RCR0635000-20F
RCR0635000-4A
Ammonia solution 20%
Ammonia solution
Formula:
NH₃ Boiling Pt: ~ 36 °C Melting Pt: ~ 0 °C Density: 0.9800 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00066650 CAS Number: 1336-21-6 UN: 3266 ADR: 8,III |
Specification Test Results
Appearance | Colorless liquid |
Assay (vs. Sulfuric Acid/Methyl Red) | 19.3 - 20.7 % (v/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...