Potassium Nitrate, 10%, pH 12 Filling Solution for Cyanide ISE, Ricca Chemical Company
Supplier: Ricca Chemical
R6319000-500A
RC631916EA 342.86 CAD
RC631916
Potassium Nitrate, 10%, pH 12 Filling Solution for Cyanide ISE, Ricca Chemical Company
Electrolytes Electrode Filling Solutions
Suitable for APHA 4500-CN F, as Reagent 3e, Potassium Nitrate Solution