Potassium nitrate 10% (w/v) in aqueous solution

Supplier: Ricca Chemical

Synonyms: Nitrate of Potash

R6317000-500A R6317000-120A
RC631716EA 308.1 CAD
RC631716 RC63174
Potassium nitrate 10% (w/v) in aqueous solution
Potassium nitrate
Formula: KNO₃
MDL Number: MFCD00011409
CAS Number: 7757-79-1
UN: 1486
ADR: 5.1,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay 10.0 ± 0.6 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR