Ammonia solution 2% (v/v)

Supplier: Ricca Chemical

R0631000-1A R0631000-4A
RC63132EA 180.12 CAD
RC63132 RCR0631000-4A
Ammonia solution 2% (v/v)
Ammonia solution
Formula: NH₃
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 0.9978 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Methyl Red) 1.90 - 2.10 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR