Potassium hydroxide 0.500 N (N/2) in methanol

Supplier: Ricca Chemical

R6261000-4A R6261000-1A
RC62611EA 483.48 CAD
RC62611 RC626132
Potassium hydroxide 0.500 N (N/2) in methanol
Potassium hydroxide
Formula: KOH
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1813
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, alcohol odor
Assay at 20 °C (traceable to NIST SRM 84) 0.5000 ± 0.0005 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR