Potassium hydroxide 0.100 N (N/10) in isopropanol (IPA)

Supplier: Ricca Chemical

R6257000-1A R6257000-4A
RC6257-32EA 238.58 CAD
RC6257-32 RC62571
Potassium hydroxide 0.100 N (N/10) in isopropanol (IPA)
Potassium hydroxide
Formula: KOH
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1813
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, Isopropanol odor
Assay at 20 °C (traceable to NIST SRM 723) 0.1000 ± 0.0005 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR