Ammonium chloride 1,15 g/L for BOD determination pH 7.20

Supplier: Ricca Chemical
Warning

Synonyms: Ammonium muriate, Salmiac

R0623000-500A R0623000-120A
RC62316EA 101.12 CAD
RC62316 RC6234
Ammonium chloride 1,15 g/L for BOD determination pH 7.20
Ammonium chloride
Formula: NH₄Cl
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0004 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
pH at 25 °C (traceable to NIST SRM 186 & 191) 7.20 ± 0.05 pH at 25.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR