Amine sulfuric acid reagent for sulfide determination (methylene blue method)

Supplier: Ricca Chemical
Danger

R0610000-500C1 R0610000-1C1
RC61016EA 303.36 CAD
RC61016 RC61032
Amine sulfuric acid reagent for sulfide determination (methylene blue method)
Amine sulfuric acid reagent
Formula: H₂O₄S
Density: 1.5000 g/cm³
Storage Temperature: Ambient
UN: 1830
ADR: 8,II

Order Now

Specification Test Results

Appearance Pale yellow to slight tan liquid, odorless
Suitability for Use Pink to Clear, colorless when Sulfide free


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR