Potassium fluoride 40% (w/v) in aqueous solution

Supplier: Ricca Chemical

R6100000-500A R6100000-1A
RC610016EA 274.92 CAD
RC610016 RC610032
Potassium fluoride 40% (w/v) in aqueous solution
Potassium fluoride
Formula: KF
MW: 58.1 g/mol
MDL Number: MFCD00011398
CAS Number: 7789-23-3
UN: 1812
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
pH at 25 °C (traceable to NIST SRM 186 & 191) 8.30 ± 0.10 pH at 25.0 °C
Specific Gravity 1.20 ± 0.01 at 25.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR