Acetate Buffer, for Iron Analysis (Phenanthroline Method), Ricca Chemical Company
Supplier: Ricca Chemical
R0060000-1A R0060000-4A
RC6032EA 199.08 CAD
RC6032 RC60-1
Acetate Buffer, for Iron Analysis (Phenanthroline Method), Ricca Chemical Company
Buffers Biological Buffers
Suitable for use in APHA 3500-Fe B as Reagent 3c