Potassium chloride saturated ∼4 M

Supplier: Ricca Chemical

R5900000-500A R5900000-1A R5900000-120A
RC590016EA 80.58 CAD
RC590016 RC590032 RC59004
Potassium chloride saturated ∼4 M
Potassium chloride
Formula: KCl
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.1700 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7

Order Now

Specification Test Results

Appearance Clear, colorless, odorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR