Potassium bromide 40 g/ 100 mL

Supplier: Ricca Chemical
Warning

R5880000-120A
RC5880-4EA 63.2 CAD
RC5880-4
Potassium bromide 40 g/ 100 mL
Potassium bromide
Formula: KBr
Boiling Pt: ~ 100 °C
Density: 1.2726 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Refractive Index 40.0 ± 0.7 g/100 mL


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR