Phosphate Buffer, pH 7, for Amperometric Determination of Residual Chlorine and Chlorine Dioxide, Ricca Chemical Company
Supplier: Ricca Chemical
R5803000-500A
R5803000-1A
RC580316EA
178.54
CAD
RC580316
RC580332
Phosphate Buffer, pH 7, for Amperometric Determination of Residual Chlorine and Chlorine Dioxide, Ricca Chemical Company
Buffers
Biological Buffers
Suitable for use in the following compendial methods:
-APHA 4500-ClO2 C, Reagent 3
-APHA 4500-Cl D, Reagent 3b
-APHA 4500-ClO2 E, Reagent 3c