Aluminium chloride

Supplier: Ricca Chemical

Synonyms: Aluminum chloride

R0570000-500A R0570000-120A
RC57016EA 567.22 CAD
RC57016 RC5704
Aluminium chloride
Aluminium chloride
Formula: AlCl₃
MDL Number: MFCD00003422
CAS Number: 7446-70-0
UN: 1726
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Refractive Index Confirmed concentration


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR