Alkaline iodide azide, Pomeroy formulation for determination of dissolved oxygen in water
Supplier: Ricca Chemical
|
Danger
|
R0535000-500A
R0535000-1A
RC53516EA
652.54
CAD
RC53516
RC53532
Alkaline iodide azide, Pomeroy formulation for determination of dissolved oxygen in water
Alkaline iodide azide, Pomeroy formulation
Boiling Pt:
~ 104 °C Density: 1.7500 g/cm³ Storage Temperature: Ambient |
UN:
1824 ADR: 8,II |
Specification Test Results
Appearance | Clear to turbid, odorless |
Free Iodine | none detectable |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...