Alkaline-iodide reagent, Winkler formulation for determination of dissolved oxygen in water

Supplier: Ricca Chemical
Danger

530-1 R0530000-500A
RC530-1EA 722.06 CAD
RC530-1 RC53016
Alkaline-iodide reagent, Winkler formulation for determination of dissolved oxygen in water
Alkaline-iodide reagent, Winkler formulation
Boiling Pt: ~ 104 °C
Density: 1.4900 g/cm³
Storage Temperature: Ambient
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless, clear to hazy, odorless
Free Iodine none detectable


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR