Methyl red 1% (w/v) in aqueous solution indicator
Supplier: Ricca Chemical
|
Danger
|
R5070000-500A
R5070000-120A
RC507016EA
202.24
CAD
RC507016
RC50704
Methyl red 1% (w/v) in aqueous solution indicator
Methyl red
Formula:
C₁₅H₁₅N₃O₂ Boiling Pt: 100 °C Melting Pt: 0.0 °C Density: 1.0050 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00002425 CAS Number: 493-52-7 |
Specification Test Results
Appearance | Clear, red, no odor |
Suitability for Use | Pink (pH 4.2) - Yellow (pH 6.2) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...