Methyl red 0,1% (w/v) in aqueous solution indicator

Supplier: Ricca Chemical
Danger

Synonyms: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid

R5060000-500A R5060000-120A
RC506016EA 99.26 CAD
RC506016 RC50604
Methyl red 0,1% (w/v) in aqueous solution indicator
Methyl red
Formula: C₁₅H₁₅N₃O₂
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0010 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00002425
CAS Number: 493-52-7

Order Now

Specification Test Results

Appearance Clear, dark red, odorless
Suitability for Use Pink (pH 4.5) - Yellow (pH 6.2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR