Alizarin red S (sodium salt) 1% (w/v) in aqueous solution indicator

Supplier: Ricca Chemical
Warning

R0500000-500A R0500000-120A
RC50016EA 135.88 CAD
RC50016 RC5004
Alizarin red S (sodium salt) 1% (w/v) in aqueous solution indicator
Alizarin Red S (sodium salt)
Formula: C₁₄H₇NaO₇S
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00013049
CAS Number: 130-22-3

Order Now

Specification Test Results

Appearance Dark, reddish-orange color, odorless
Suitability for Use Yellow (pH 4.6) - Pink (pH 6.0)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR